Ice buckets, round, with lid, Magic Touch™
Supplier: Bel-Art
BELAM188482002EA 138 EUR
BELAM188482002 BELAM188482201 BELAM188482202 BELAM188484001 BELAM188484201 BELAM188482001 BELAM188484002
Ice buckets, round, with lid, Magic Touch™
Expanded urethane.
  • Temperature range: -196° to +100 °C
  • Lid can be cut/drilled to make openings for thermometers, cooling coils, flasks etc.
  • Holds Magic Touch round floating racks
Superior for retaining hot or cold, Magic Touch insulated ice buckets are durable, stackable and lightweight. Store and transport temperature sensitive samples in ice, alcohol slurries, water baths and more. Use with ice, dry ice, dry ice alcohol slurries, liquid nitrogen. Expanded polymer foam is chemical resistant, impervious to moisture and odors, and unbreakable under normal use conditions.
Ordering information: Magic Touch 2™ ice buckets are available in 2,5 litre and 4,0 litre sizes, each in two colours; black and blue.