Digital thermometer, Traceable
620-2021EA 63.9 EUR
VWRU61161-274 VWRI620-2021
Digital thermometer, Traceable
This thermometer is designed to monitor temperatures in food, soil, cuvettes, water baths, wastewater, or incubators around the clock. Both a high and low alarm may be programmed in 1° increments: When temperature rises above or falls below set points, a 1 minute audible alarm signals an 'out of range' condition.
  • Countdown timer set in 1 minute increments
  • Audible alarm for temperature
  • Maximum/minimum reading
External stainless steel probe (L×Ø: 184×3,6 mm) with 90 cm cable
Delivery information: Supplied with a flip-open stand for bench use, a probe, a AAA battery, and a Traceable® certificate.
Certifications: Conforms to NIST standards, ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with flip-open stand for bench use, probe, AAA battery and Traceable certificate of calibration.