Safety canister, disposal units
Supplier: SCAT Europe
590-0546EA 155 EUR
590-0546
Safety canister, disposal units
Safety Cans
HDPE.
These ready to use sets provide complete solutions for the disposal of solvent waste. They offer more protection for people and the environment whilst reducing the amount of solvents used.
Canisters with SafetyWasteCap and filter.
Safety caps for almost every conventional disposal container.
Exhaust filter (size S) absorbs 99% of particulate materials evaporating from the solvents.
Canisters, electroconductive with level control and funnel.
Electroconductive sets are suitable for use in explosion-proof areas. Level control is integrated into the canister, a visual display warns users when a container may overflow. Funnel includes sieve that can be removed for cleaning purposes.
Canisters, electroconductive with funnel with cap, sieve and drain tray.
Drain trays essential for systems without level control. Avoid spilling solvents on the floor if the container overflows or if there is an accident during filling. SCAT drain trays have a much lower weight than stainless steel trays.
Canisters with funnel with cap and sieve.
Funnels are suitable for all types of chemicals. Screw caps rotate freely for easier screwing on and off the funnels. Vials can be disposed of completely with these sets.