Mechanical Balances, Field Test
Supplier: Ohaus
611-5196EA 1490 EUR
611-5196
Mechanical Balances, Field Test
Balances and Scales
The Field Test Scale provides accurate weighing of a wide variety of heavy materials, even under the most adverse conditions. The Field Test Scale is ruggedly built and designed for field applications where accurate measurements are essential.