8-Quinolinol ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
26123.237EA
158
EUR
26123.237
8-Quinolinol ≥99.0%, AnalaR NORMAPUR® analytical reagent
8-Quinolinol
Formula:
C₉H₇NO MW: 145,16 g/mol Boiling Pt: 267 °C (1013 hPa) Melting Pt: 71,2 °C Density: 1,03 g/cm³ (20 °C) |
MDL Number:
MFCD00006807 CAS Number: 148-24-3 Merck Index: 13,04869 |
Specification Test Results
Assay | Min. 99.0 % |
Melting point | 72.5 - 74.0 °C |
Ignition residue (SO4) | Max. 0.02 % |
Insolubility in acetone | Max. 50 ppm |
Insolubility in HCl 2 N | Max. 100 ppm |
Insolubility in NaOH 1 N (± 80°C) | Max. 100 ppm |
Cl (Chloride) | Max. 20 ppm |
SO4 (Sulphate) | Max. 0.02 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...