Automated slide stainer, AutoStainer XL, ST5010
Supplier: Leica
720-1878
Automated slide stainer, AutoStainer XL, ST5010
Slide Stainers
Programmable stainer for high flexibility and reliability in histology and cytology featuring walk-away convenience. As soon as the staining procedure is completed, an innovative robot arm transfers the slide racks from the stainer to the coverslipper.
  • Incubation time and agitation flexible
  • Air filter vent with charcoal filter to retain solvent fumes
  • Communication and remote alarm port
Certifications: In compliance with UL/c-UL and CSA regulations.
Delivery information: Supplied an accessory kit consisting of 22 reagent vessels with lids, 5 wash vessels, 10 slide racks, 2 slotted lids for reagent vessels, jumper cable, tubing band clamp, elbow hose, V-filter for inlet hose, jack connector (remote alarm), activated carbon filter, fume cover, wax collection tray (heater), programme pad, inlet hose with, drain hose, set of power cords, EC declaration of conformity, instructions for use and a reference manual.