Blanks for jacketed vessels, round bottom, DURAN®
Supplier: DWK Life Sciences
SCOT210276275EA 143.23 EUR
SCOT210276275 SCOT210275373 SCOT210274274 SCOT210274771 SCOT210276678 SCOT210275579 SCOT210273775 SCOT210275879
Blanks for jacketed vessels, round bottom, DURAN®
Reaction Vessels Reaction Vessels, Cylindrical Shape
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Round bottom cylinders.