Microtitre plate shaker incubator, SI505
Supplier: Cole Parmer
444-0692EA 4060 EUR
STUASI505
Microtitre plate shaker incubator, SI505
The SI505 has been designed specifically to combine a bench top laboratory incubator with the specific mixing action required for microtitre plates. Shaking is ideal for mixing microtitre plates, combining a high speed action between 250 and 1250rpm with a tight orbit of 1,5mm. This enables even the smallest of vessels to adequately mix, be it well plates or microtubes.
  • Ideal shaking action for microtitre plates
  • Combined incubator shaker
  • Optimised performance to minimise sample evaporation
  • Digital display for temperature and speed
  • Integrated timer
  • Retractable stainless steel platform for easy loading and unloading
  • BioCote® antimicrobial protection
The SI505 has independent control of speed and temperature to avoid accidental temperature adjustment. Speed is microprocessor controller and set via the digital LED control panel. The unit also incorporates a versatile timer, which can be set from one second to nine days, once the timer has run down, an alarm will sound and the shaking action will cease, for safety reasons the incubator will continue to run.
The temperature of the incubator can be set, via the digital LED control panel, between ambient temperature +7 °C and 60 °C. Careful control of the air distribution within the incubator ensures temperature uniformity throughout the sample chamber.
To minimize sample evaporation the SI505 is supplied with stainless steel water trays, these are strategically placed within the unit to maintain a humidity of up to 80%, thus dramatically reducing sample evaporation over a 24 hour period. During operation the platform is securely locked in place, but can be easily released when required.
The platform will comfortably and securely accommodate up to four microtitre plates on the non slip mat. Or sample tube racks are available which will accommodate all common tube sizes. The SI505 also features a USB connection which allows the incubator to be connected to a PC for long term monitoring of the incubator temperature, over night of weekends for example.
Certifications: IP protection class according to DIN EN 60529: IP 31.
Delivery information: Supplied with stainless steel water trays. IQ/OQ documentation is available as an optional accessory if required.