Container, conical, for snap-on lids
Supplier: Gosselin
GOSSPC200A-05EA 184 EUR
GOSSPC200A-05 GOSSPC200A-07 215-0791 GOSSPC200A-11 GOSSPC200A-03
Container, conical, for snap-on lids
Sample Containers General Purpose Containers
Made of PP, white or natural.
  • Good chemical resistance
  • Good temperature resistance
  • Available with or without scap cap
Ideal for collecting, storing and transporting samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904