Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Cryogenic Labels"
https://ru-bot.vwr.com/stibo/smallweb/std.lang.all/50/84/18945084.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   PolarSafe Dots are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware. These dots w...
Catalog Number: (479-3291)
Supplier:  Nalgene
Description:   Cloth labels for use at ultra-low temperatures, ballpoint pen compatible.
UOM:  1 * 200 items
Supplier:  Bel-Art
Description:   The strong, permanent adhesive on these labels withstands cold storage temperatures and maintains adhesion to glass, plastic, cardboard and more, even...
Catalog Number: (HEATHEALCRY-1700)
Supplier:  HEATHROW
Description:   These labels are temperature resistant -196 to +150 °C and are chemically inert.
UOM:  1 * 1.700 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4  of 4