Alu rings with hooks for conical joints
Supplier: Lenz
LENZ01102519EA 55.6 EUR
LENZ01102519 LENZ01102545 LENZ01102529 201-7425
Alu rings with hooks for conical joints
Clamps NS Joint Clamps
These alu rings with hooks are for cones and sockets to be used with springs for the safe connection of conical ground joints. No breakage of glass hoods.