Egomat® Aqua mats are made from premium EPDM rubber. The mats are designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave. Also suitable for outdoor use.
- Upper side with cones
- Autoclavable, withstands 600 °C (welding)
- Resistant against some alkaline liquids, acids and alcohols
- Frequent washes will not affect the state of the mat; no curling up, swelling or expanding
- Anti-fatigue and ergonomic benefits due to durability and unique moulded design
- Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Green, red or grey
Certifications: Meets EN ISO/USDA/CFIA standards