These state of the art down-flow cabinets, provide an ultimate cleanliness Class 100 work area where the highest safety for the products is achieved. The internal design, the air flow aerodynamics and monitoring, the special H14 fi lter with micromesh downstream equalising plenum, guarantees the highest performances at the most stringent safety levels and operator comfort. The special sloped front design and the sliding sash ensure comfortable working position and, when needed, a very easy opening and closing of the front window.
- Tempered safety glass side walls for the best visibility and lightning
- Unique x/y sliding sash window with three positions
- Centrifugal motorblower
- Removable, perforated work surface and back wall of internal chamber made from grade 3 stainless steel
- Soft touch keys on control panel provide control of lighting, motorblower, UV light, solenoid valve and power point
- Visual and acoustic alarms for fan failure
This unit works on a recirculating mode with air being filtered by an HEPA H14 filter with 99,999% efficiency on 0,3 µ particles, before entering the work area at a speed of 0,45 m/s; this ensures a Class 100 environment throughout the work area. Air coming through the front aperture is then mixed with the downfl ow air, without affecting the product, and recirculated. 70% of the air is returned to the working area through the main HEPA fi lter and 30% is exhausted into the environment through an exhaust HEPA fi lter. This ensures double protection both for the product and for the operator even if this cabinet should not be used for handling pathogens.
Rigid steel frame (paint finished) with laminated glass front sliding window and side panels, the working surface is made with AISI 304 stainless steel
Certifications: CE marked. All BIO-Cabinets products conform to strict International Standards ; manufactured and designed to comply with AS1386.5.
Delivery information: Delivery prices, installation and commissioning are available on request.