Carboys, narrow neck, PFA, Chemware®
Supplier: Saint-Gobain
VERND1069730EA 16320 EUR
VERND1069730
Carboys, narrow neck, PFA, Chemware®
PFA, translucent.
  • Chemically inert and ultra-pure; meets USP Class VI criteria
  • Lowest metal ion content of all fluoropolymers
  • Ideal for storage during intermediate phases of pharmaceutical and semiconductor processing
Leak-proof, unbreakable PFA carboys are used for storage of any corrosive chemicals, including hydrochloric, nitric and perchloric acids. They are also ideal for storage of HPLC mobile phase. PFA carboys reduce the risk associated with broken glass and solvent spills. They have an inert fluoropolymer plug-type seal and a PTFE cap.