VWR®, Carboys, Wide Neck, BPA-free
Supplier: Avantor
216-0612EA 108 EUR
VWRI216-0612
VWR®, Carboys, Wide Neck, BPA-free
BPA-free, clear.
  • Versatile Cap technology provides the flexibility of interchangeable connectors
  • Extra wide mouth design for easy filling and cleaning
  • Smart Barb technology provides interchangeable spigot options
This BPA-free carboy is an advanced fluid handling system developed with the end user in mind. The ergonomically-designed carboy body, coupled with our VWR® Versatile Cap technology, provides the user with something that is easy to use and easy to clean. It is made from a material that is heat stable. BPA-free carboys are ideal for aqueous solutions, and are used for most life science buffer solutions. The carboy body includes design features that allow the user to easily hold and manipulate during lifting, filling and pouring, while Versatile Cap technology provides interchangeable, leakproof top connections. Features include a set of handles at both the top and bottom, easy to read embossed graduation marks, and moulded carboy material type indentification mark. 20 and 40 litre carboys are also available with an extra wide mouth for easier filling and cleaning.