PCR racks for tubes and plates
Supplier: Eppendorf
713-0004EA 164 EUR
713-0004
PCR racks for tubes and plates
Racks Tube Racks
PP.
  • For 0,1, 0,2 and 0,5 ml tubes, as well as 96-well plates
  • Stackable
  • Can be stored down to −90 °C
  • Autoclavable (121 °C, 20 min)
Rack for handling and storing tubes and plates.