Half masks, 3000 series
Supplier: 3M
MMMA3100SEA 11.8 EUR
MMMA3100S
Half masks, 3000 series
The 3000 series half-mask is a compact and lightweight mask that offers an economical solution for a range of applications, securing high levels of working protection and comfort.
  • Low profile design allows a wide field of vision
  • Lightweight makes long period of wear bearable
  • Soft, elastomeric face-fit secures lasting comfort
  • Unique, easy to don/doff strap system
It moulds itself softly and securely to the contours of the face without blocking the view. The user can quickly snap-on the relevant gas or particulate option and keep the breathing at ease.
Mask can be combined with a variety of cartridges or respiratory filters.
Certifications: EN 140
Ordering information: Cartidges and respiratory filters are ordered seperately.