Hand pads, general purpose, 7447, Scotch-Brite™
Supplier: 3M
115-1751EA 47.6 EUR
MMMA07447
Hand pads, general purpose, 7447, Scotch-Brite™
This hand pad is most universally used in a variety of applications. It can be used for cleaning, finishing, graining, denibbing and defuzzing. The pad may be used by hand, with a hand pad block, or on an in-line sander.
  • Standard for surface conditioning worldwide
  • Clean, blend, finish, and deburr faster, easier, and with better results than conventional abrasives
  • Provides similar performance as steel wool grade 1
Abrasive mineral: Very fine grade aluminum oxide.