Environmental test chambers, MKT series
Supplier: Binder
BIND9020-0082EA 0 EUR
BIND9020-0082 BIND9020-0196
Environmental test chambers, MKT series
Testing Chambers Environmental Test Chambers
The MKT series of dynamic climate chambers has been specially developed as precision heating and cooling chambers for industrial material testing and environment simulation applications. The units are ideal for rapid temperature changes with a low temperature range down to –70 °C and have a higher capacity which exceeds the capability of normal testing chambers. The stainless steel inner chamber has door heating, a heated viewing window with LED interior lighting and programmable condensation protection for the test materials.
  • MCS controller can activate four zero-voltage relay contacts
  • APT line™ preheating chamber technology and air conditioning technology provides spatial temperature for total working area
  • User friendly colour LCD screen and easy to read menu guide, real time clock and integrated electronic chart recorder
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Adjustable ramp function, PT 100 temperature sensor and CFC-free refrigerant R-404A
Certifications: IP protection class according to DIN EN 60529: IP 20. Conform to DIN EN 60068-2-1: 2008-01, DIN EN 60068-3-5: 2002-12, and MIL STD 810:2000-01
Delivery information: Supplied with one stainless steel shelf, Binder test certificate, cooling with compressor cooling unit, evaporator, CFC-free refrigerant R-404A, four castors, two with brakes, thermal insulation, PT 100 temperature sensor and APT-COM™ 3 Basic edition communication software. Units have an Ethernet interface, MKT 115 and MKT 240 have 50 mm Ø access port with silicone plug on the left , while MKT 720 has 2× 80 mm access ports on the left and right. A wide range of accessories are available, calibration and validation is possible, for details please visit www.vwr.com or contact your local Avantor sales office. Please note all MKT units require a three phase supply.