Isopropyl acetate, Supelco®
M-Clarity™ quality level = MQ100
90871-10ML-FEA 156 EUR
90871-10ML-F 90871-1ML-F 90871-5ML-F
Isopropyl acetate, Supelco®
Isopropyl acetate
Formula: C₅H₁₀O₂
MW: 102,13 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0,8732 g/cm³ (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00008877
CAS Number: 108-21-4
UN: 1220
ADR: 3,II
Merck Index: 13,05225