Potassium bromate 1/60 mol concentrated aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
4667.EA 41.54 EUR
4667.
Potassium bromate 1/60 mol concentrated aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Potassium bromate
Formula: KBrO₃
MW: 167 g/mol
Melting Pt: 350°C
MDL Number: MFCD00011359
UN: 1484
ADR: 5.1,II
Order Now
Specification Test Results
Titer (N) 0.0998-0.1002