Overalls, KLEENGUARD* A50
Supplier: Kimberly-Clark
KleenGuard®
113-8730EA 682 EUR
113-8730 113-8731 113-8729 113-8727 113-8728 113-8725 113-8726 113-8734 113-8724 113-8735 113-8732 113-8733
Overalls, KLEENGUARD* A50
Disposable Garments
Disposable overall with hood that offers protection against chemical splash and particles. The hood is designed for mask use and freedom of movement, the strong triple stitched seams help to protect against tearing.
  • Anti-static according to EN 1149-1 for critical areas
  • Low lint fabric and internal seams reduce the risk of fibre contamination
  • Silicone-free, ideal for paint spraying
  • Two-way zip for quick easy access to work clothes
The clothing is made of durable and breathable SMS fabric and therefore suitable for chemical, pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying.
Colour: White or Blue
Certifications: Type 5/6, EN 1073-2, EN 1149-1; Cat. III