Graduated pipettes for tissue culture, total delivery
Supplier: Brand
BRND27164EA 58.2 EUR
BRND27164
Graduated pipettes for tissue culture, total delivery
AR-Glas®, tissue culture short type.
  • Calibrated to deliver (TD, Ex)
  • Graduations and inscriptions in ETERNA amber stain
  • Ring marks at major graduations
Length: 230 mm