Microplate readers, Multiskan™ GO
Supplier: Thermo Scientific
736-0559EA 10930 EUR
LABL51119200
Microplate readers, Multiskan™ GO
The Multiskan™ GO is a compact, reliable, absorbance spectrophotometer microplate reader available in two instrument models, with or without a cuvette port. The instrument can be operated as a stand-alone unit using straightforward internal software or by using Thermo Scientific SkanIt™ software for PC control. Both the internal and PC software are available in multiple language versions, including English, French, German, Spanish, Portuguese, Russian, Chinese and Japanese. A broad wavelength range, including UV, makes this microplate reader ideal for a wide range of photometric applications, such as nucleic acid and protein analysis, enzyme assays, cytotoxicity and cell proliferation assays as well as apoptosis assays. The cuvette option provides a quick and easy way of measuring when there are only a couple of samples to be analysed.
  • Freely selectable wavelengths by the monochromator for the demands of various assays
  • Fast plate measurement and a full spectrum of a sample in less than 10 seconds
  • Both microplate and cuvette reading capability available
  • Shaking and incubation up to 45 °C for temperature-critical applications
  • Robotic compatibility for high throughput environments
The SkanIt™ software package supplied with the instruments provides easy assay optimisation, flexible data handling and convenient report formatting. The software also has a special remote control interface, which enables easy integration with automated systems.
µDrop™ Plate is a quick and easy tool for DNA and RNA assays using photometric measurement and sample volumes down to 2 µl. The fixed light path of the µDrop™ Plate allows direct calculation of the nucleic acid concentrations of the samples.
dsDNA detection range from a few micrograms to a few milligrams per millilitre
Sixteen sample positions for quick and easy measurement of sample volumes down to 2 µl
Compatible with an eight channel pipette for easy sample addition
Quick and easy to wipe off the samples in serial measurements
Format flexibility with measurement slot for a rectangular cuvette (10 mm path length)
Compatible with Multiskan™ GO and Varioskan™ Flash