Air sampler, AirPort MD8
Supplier: Sartorius
SART16757EA 4620 EUR
SART16757
Air sampler, AirPort MD8
AirPort MD8 is a battery-powered portable air sampler.
  • Battery power level clearly indicated for constant performance
  • Ergonomic design and easy cleaning
  • Adjustable flow and volume sample
  • Last parameters stored after automatic shut-off
  • User-friendly prompting in four languages; English, French, German, Spanish
Suitable for the pharmaceutical industry, biotechnologies, food and beverage applications, hospitals environment and work safety.
Ordering information: Includes an adapter for gelatin filter disposables and battery charger.