Mortars with pestles
Supplier: Brand
BRND110705EA 0 EUR
BRND110705
Mortars with pestles
Mortars and Pestles Mortars with Pestles
Melamine formaldehyde, white.
  • Resistant to boiling and autoclavable (121 °C)
  • With foot and spout