You Searched For: Liquid Process Filters
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: These filters are made from 100% PEEK.
Supplier: Avantor
Description: The Last Dropâ„¢ eluent filter utilises a flat element, which sits parallel to the bottom of the reservoir, allowing the filter to draw all but the last...
Supplier: Avantor
Description: These solvent inlet filters are used at the low pressure inlet side of the pump to help protect check valves, injector and column from particulate con...
Supplier: ROBU GLASFILTER
Description: These filters are suitable for degassing and filtration of solvents e.g. in chromatography.
Supplier: Bohlender
Description: These PTFE suction filters consist of a receiver with a female thread and an easily exchangeable frit. They are ideal prefilters in front of pump syst...
Supplier: ENTEGRIS INC
Description: PUMPGARDâ„¢II filters protect chemicals and final filters against damage from broken silicone wafer pieces.
Catalog Number: (590-0221)
Supplier: Bohlender
Description: Suction Filter with PTFE membrane.
UOM: 1 * 5 items
Catalog Number: (ROBU80052)
Supplier: ROBU GLASFILTER
Description: This solvent filter is made of pure borosilicate glass.
UOM: 1 * 1 items
Catalog Number: (590-0201)
Supplier: Bohlender
Description: Filter candle made of porous PE.
UOM: 1 * 10 items
Catalog Number: (VIVAH2O-CUF)
Supplier: Sartorius
Description: The arium® CellPlus is a sterile packed, point-of-use ultrafilter for efficient removal of endotoxins, RNase, DNase, microorganisms and particles. Ret...
UOM: 1 * 1 items
Supplier: IDEX HEALTH&SCIENCE LLC(RHEODY
Description: Inlet filters are used to extract particles from the sample between the injector and the column.
Supplier: PerkinElmer
Description: Solvent reservoir sparger, Stainless steel, Pore size: 40 µm
Catalog Number: (REST26431)
Supplier: Restek
Description: This solvent filter is made up of borosilicate glass and is used to prevent the particulates and microbial growth in the solvents from entering the sy...
UOM: 1 * 1 items
Supplier: Agilent
Description: It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
Supplier: Restek
Description: These purge and trap spargers are designed for use with Tekmar purge-and-trap instruments only and manufactured to tight tolerances to ensure a leak-t...
Catalog Number: (REST25008)
Supplier: Restek
Description: This slip-on inlet filter is made of 316 stainless steel tip with a tefzel collar seals to a corrosion-resistant 316 stainless steel filter element.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom