You Searched For: Spectrum Lamps
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Spectrum lamp, Tungsten halogen lamp
Supplier: Hach
Description: Spectrum lamp, Deuterium lamp
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Spectrum lamp, Rare gas, Pen-Ray®, Xenon lamp
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Spectrum lamp, Zinc, axial leads, 203 mm
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Spectrum lamp, Pen-Ray®, Calibration lamp, CP series, 254 nm, 47 mm
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The BioLite MultiSpectral Source uses a 150 W quartz halogen or 150 W xenon light source with fibre optics to tightly control the output spe...
Supplier: DIDALAB
Description: Nine pin socket spectral lamps with dimensions (H×Ø) 107×21 mm, operated with 1 Amp fixed current.
Catalog Number: (ZEIS3901580000000)
Supplier: ZEISS
Description: Spectrum lamp, Tungsten lamp filament, 6 V, 30 W
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom