You Searched For: Mercury Lamps
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Pen-Ray®, Mercury lamp, quartz, 11SC-2, 254 nm, with standard connector, For: Analysis, photoreaction, calibration and restricted access, Height: 82,6 mm
Catalog Number: (28-4042-25)
Supplier: Cytiva
Description: Mercury lamp and housing, UPC-900
UOM: 1 * 1 items
Catalog Number: (ZEIS0000001313162)
Supplier: ZEISS
Description: This mercury short arc reflector lamp is specially designed for X-Cite 120 illuminator system. The average lifetime performance of this lamp module is...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 3 of 3
no targeter for Bottom