Multicoated multiwell plates, Corning® BioCoat™
Supplier: Corning
Corning® BioCoat®
734-1043EA 0 EUR
734-1043
Multicoated multiwell plates, Corning® BioCoat™
Microplates Cell Culture Plates
PS, clear, TC-treated, flat bottom. Each Corning® BioCoat™ 6-well Multiwell variety pack contains five clear TC-treated polystyrene plates with a selection of different extracellular matrix proteins and attachment factors.