You Searched For: Weights
56  results were found
SearchResultCount:"56"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Test weights in either stainless steel or aluminium.
Supplier: VWR Collection
Description: Stainless steel. Wire weights up to 500 mg in cobalt alloy.
Supplier: VWR Collection
Description: Weights made from anti-magnetic, polished stainless steel. Suitable for medium resolution, precision electronic balances. Weights of more than 20 g ha...
Supplier: VWR Collection
Description: Rectangular weights made from cast iron, permanently magnetised.
Supplier: VWR Collection
Description: Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.
Supplier: VWR Collection
Description: Weights from 1 to 50 g are monobloc Weights, with larger Weights having an adjusting cavity.
Supplier: VWR Collection
Description: Weights made from anti-magnetic, polished stainless steel. Suitable for medium resolution precision electronic balances. Weights have adjustment cavit...
Supplier: VWR Collection
Description: Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Supplier: VWR Collection
Description: Weights made from solid, anti-magnetic, polished stainless steel.
Supplier: VWR Collection
Description: An economical range of calibration sets, with only three stainless steel weights, to meet routine calibration requirements of precision and analytical...
Supplier: VWR Collection
Description: Weights made from solid, anti-magnetic, polished stainless steel. Suitable for high resolution electronic balances.
Supplier: VWR Collection
Description: Rectangular weights made from cast iron (density 7,2 g/cm³).
Supplier: VWR Collection
Description: Brass weights that are ideal for schools introducing the concept of weights and forces.
Supplier: VWR Collection
Description: Individual E1 OIML class weights are intended to ensure traceability between the national mass standards (with values derived from the International P...
Supplier: Avantor Fluid Handling
Description: Keep your samples flowing and tubing from curling.
Catalog Number: (611-3815)
Supplier: VWR Collection
Description: Weight, Calibration, VWR®, Class: E1, Weight: 1000 g, Stainless steel
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 56
no targeter for Bottom