Calendar up to the year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch and Danish), 12-/24-hour selectable time with snooze alarm, moon phase and animation weather forecast.
- Comfortable index icons
- Links up to three remote modules (maximum distance 30 metres)
- Dual display to show the temperature of the main unit and one external sensor
Calendar up to the year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch and Danish), a 12/24-hour selectable time with snooze alarm, moon phase, and animation weather forecast.
- Comfortable index icons
- Links up to three remote modules (maximum distance 30 metres)
- Dual display to show the temperature of the main unit and one external sensor at a time
W×H: 117×193 mm
Compliant with RoHS.
Certifications: Compliant with RoHS.
Delivery information: Supplied with batteries.