Oil baths, ONE Series
Supplier: Memmert
462-3576EA 0 EUR
462-3576
Oil baths, ONE Series
Baths Oil Baths
Oil baths with corrosion resistant stainless steel tank and outer case. Units have two PT100 sensors, the heating elements are in the sides and on the bottom of the tank giving better temperature uniformity.
  • PID microprocessor controller with an integrated self-diagnostic system and digital LED display for all set parameters and alarm values, with 0,1 °C resolution (for temperatures below 99,9 °C).
  • Programmable digital timer, 1 min. to 999 h with four functions; on, delayed on, hold, hold set temperature dependent with defined holding time
  • Acoustic and visual alarm for low liquid level; heating is automatically switched off at the end of the programmel
  • Safety features include an independent adjustable temperature limiter, TWB protection class 2
Ordering information: All models are supplied without racks, ring sets or lid. Please order required accessories separately.
Caution: * RT= Ambient