You Searched For: Baths
79  results were found
SearchResultCount:"79"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Made from stainless steel or polycarbonate. Ideal for use with VWR® immersion thermostats. For use where the durability of stainless steel or the tran...
Sale
Supplier: VWR Collection
Description: Stainless steel baths with user friendly MX temperature controller with three control buttons and on-screen prompts.
Catalog Number: (462-0214)
Supplier: VWR Collection
Description: Stainless steel bath with user friendly MX temperature controller, has three control buttons and on-screen prompts. Specifically designed for producti...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Transparent polycarbonate baths keep samples visible, units feature user friendly MX temperature controller with three control buttons and on-screen p...
Supplier: VWR Collection
Description: Linear shaking water baths with stainless steel tank and simple, intuitive user interface. It is quick and easy to fit alternative shaking trays.
Supplier: VWR Collection
Description: Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 litre models feature ...
Catalog Number: (720-2423)
Supplier: VWR Collection
Description: The reliable W20 histology water bath is ideal for removing wrinkles and distortions in paraffin wax sections which are created during sectioning. The...
UOM: 1 * 1 items
Supplier: JULABO GmbH
Description: Unstirred water bath, CORIO™, B19, Stainless steel, Capacity: 14 - 19 L, Temperature range: ≤150 °C, Bath W×D×H: 300×500×150 mm, W×D×H: 370×580×200 mm
Supplier: JULABO GmbH
Description: Unstirred water bath, CORIO™, BT9, Capacity: 6 - 9 L, Temperature range: ≤100 °C, Bath W×D×H: 230×150×150 mm
Supplier: GRANT INSTRUMENTS LTD.
Description: High quality, digital unstirred water bath range with choice of models from 2 to 34 litres and a dual bath with 5 and 12 litres. Dual baths are ideal ...
Supplier: JULABO GmbH
Description: Stainless steel shaking water baths are suitable for unsupervised continuous operation. The bright, LED Multi-Display showing up to six temperature va...
Supplier: JULABO GmbH
Description: PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use...
Supplier: GRANT INSTRUMENTS LTD.
Description: Digital unstirred water bath range with choice of models from 5 to 18 litres. All units have stainless steel tanks and durable painted steel cases, of...
Supplier: GRANT INSTRUMENTS LTD.
Description: The Aqua Pro series of shaking water baths offers two bath models with exceptional temperature performance and usability. The baths are ideal for basi...
Catalog Number: (462-0519)
Supplier: GRANT INSTRUMENTS LTD.
Description: The OLS shaking water bath offers exceptional temperature performance and usability. The bath is ideal for basic as well as more demanding application...
UOM: 1 * 1 items
Catalog Number: (JULA9461701)
Supplier: JULABO GmbH
Description: Open bath circulator with high quality bath tanks made of transparent plastic or stainless steel.
UOM: 1 * 1 items
New Product
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 79
no targeter for Bottom