You Searched For: Vacuum Valves
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ENTEGRIS INC
Description: Two- or three-way diaphragm valves, ¹/₄" orifice, normally closed.
Supplier: VACUUBRAND GMBH + CO
Description: High vacuum, helium leak-tested right angle valves VE have stainless steel bellows which seals the drive mechanism without any gap on the vacuum side.
Supplier: VACUUBRAND GMBH + CO
Description: The VS, VS C and VS B series valves are butterfly-type valves which offers very good conductance. The VS C series features a valve seal made of perflu...
Supplier: VACUUBRAND GMBH + CO
Description: The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popula...
Supplier: Welch by Gardner Denver
Description: These ball check valves have a PTFE hollow ball and viton seals.
Supplier: VACUUBRAND GMBH + CO
Description: VM series valves are hand operated diaphragm valves. They are used as isolation valves, air-admittance valves or manual-control valves.
Supplier: VACUUBRAND GMBH + CO
Description: Valves of the VK series are sturdy isolation devices for the rough and fine vacuum range. The VKE series uses stainless steel for the metal parts and ...
Supplier: Welch by Gardner Denver
Description: These manually operated butterfly valves are made of stainless steel with viton seal.
Supplier: Welch by Gardner Denver
Description: These high vacuum through valves feature a very long life bellows-seal made from stainless steel.
Supplier: Welch by Gardner Denver
Description: These high vacuum angle valves feature a very long life bellows-seal made from stainless steel.
Supplier: VACUUBRAND GMBH + CO
Description: These straight-through valves with electromagnetic (solenoid) drive are used for operating cycles with short intervals and it is often used as vacuum ...
Supplier: VACUUBRAND GMBH + CO
Description: Gas inlet valves are small hand operated valves for admitting gases into a vacuum system.
Supplier: Welch by Gardner Denver
Description: These fine vacuum ball valves feature a small flange made of nickle plated aluminium.
Supplier: Restek
Description: Ball valves have a floating ball to assist sealing and to reduce operating torque, and dual seats to provide leak-tight bidirectional sealing. They ar...
Catalog Number: (229-7485)
Supplier: SEMADENI
Description: Three way ball valve with PP body and PTFE tips with total length of 65 mm.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom