You Searched For: Vacuum Controllers
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (181-0332)
Supplier: VACUUBRAND GMBH + CO
Description: The CVC 3000 detect compact vacuum controller complete with integral vacuum valve and sensor is ready to use either as a bench top or stand mounted un...
UOM: 1 * 1 items
Safer Human and Environmental Health
Catalog Number: (131-0000)
Supplier: VACUUBRAND GMBH + CO
Description: The VACUU·SELECT controller is an intuitive, easy-to-use vacuum controller with a touch screen display suitable for all laboratory vacuum processes. F...
UOM: 1 * 1 items
Catalog Number: (KNFAVC900)
Supplier: KNF
Description: Vacuum control unit to control the vacuum applications, like rotary evaporation, distillation, multi-user vacuum system.
UOM: 1 * 1 items
Catalog Number: (181-0581)
Supplier: Welch by Gardner Denver
Description: Precise vacuum control for evaporation and distillation of solvents and chemicals.
UOM: 1 * 1 items
Supplier: VACUUBRAND GMBH + CO
Description: The VACUU·SELECT complete controller is ready for use with existing vacuum sources like pumps or vacuum networks.
Supplier: VACUUBRAND GMBH + CO
Description: VACUU·SELECT fine vacuum control packages provide an easy-to-use vacuum control option for work between 1 mbar and 10-3 mbar, such as freez...
Catalog Number: (VACU20683180)
Supplier: VACUUBRAND GMBH + CO
Description: The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popula...
UOM: 1 * 1 items
Catalog Number: (181-0140)
Supplier: BUCHI
Description: V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustme...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom