You Searched For: Transilluminators
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: All VWR® transilluminators slide out for easy access to provide very even illumination. Those for use with the VWR® Imager system also have variable i...
Catalog Number: (730-1488)
Supplier: VWR Collection
Description: The VWR® benchtop transilluminator for stand alone imaging systems is designed for ease of use.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. ...
Supplier: Syngene (a division of the Synoptics group)
Description: UV Transilluminator, Syngene, GX-2020SL-E, Wavelength: 254/365 nm, Filter size: 200×200 mm
Catalog Number: (SYNOGX-ULTRABRIGHT)
Supplier: Syngene (a division of the Synoptics group)
Description: Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. ...
UOM: 1 * 1 items
Catalog Number: (SYNOGEULTRASLIMLED)
Supplier: Syngene (a division of the Synoptics group)
Description: UltraSlim-LED transilluminators for superior gel illumination.
UOM: 1 * 1 items
Catalog Number: (733-2310)
Supplier: Syngene (a division of the Synoptics group)
Description: UV Transilluminator, Syngene, GE-2020M-E, Wavelength: 302 nm, Filter size: 200×200 mm
UOM: 1 * 1 items
Catalog Number: (730-1002)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: White Light transilluminators offer uniform Light diffusion for autoradiographs, negatives, X-ray film, film positives, microtitre plates and gels.
UOM: 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: These High Performance UV transilluminators, with four 25 W tubes, provide 100 W of high intensity and uniform UV illumination for high fluorescence a...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The Visi-Blueâ„¢ converter plate is simply placed on top of a transilluminator equipped with mid-range (302 nm) UV to convert the UV to 460 to 470 nm bl...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The specially designed converter plate converts UV radiation from the transilluminator using a white light diffuser to enable Coomassie® Blue or silve...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: These UV/white light transilluminators house white light and UV working areas side-by-side. White light can be used for viewing Coomassie Blue stained...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: All Benchtop transilluminators are compact, lightweight and offer an economical alternative to the High Performance series transilluminators. With the...
Catalog Number: (UVPA38-0325-01)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV Transilluminator
UOM: 1 * 1 items
Catalog Number: (733-2005)
Supplier: EDVOTEK EUROPE LTD
Description: The TruBlu™ blue light transilluminator is ideal for viewing DNA gels stained with SYBR® Safe, thus eliminating the need for UV light or ethidium brom...
UOM: 1 * 1 items
Catalog Number: (EMBIVL-1200)
Supplier: EMBITEC
Description: Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
UOM: 1 * 1 SET
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom