You Searched For: Turbidity Meters
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: TIR 210 is a compact turbidity meter designed for fast and precise on-site testing.
Supplier: Hach
Description: Tungsten or LED light sources, as well as proven optics, make these instruments more reliable to deliver accurate and stable measurements.
Supplier: WTW
Description: The Turb 355 provide the user with the choice to perform nephelometric measurements at 90° scattered light, either with an infrared or tungsten white ...
Supplier: WTW
Description: The Turb 430, with lab option via LabStation, include drinking water testing, environmental monitoring, laboratory reference measurements and process ...
Supplier: Hach
Description: These laser turbidimeters feature performant optical technology. Radio-frequency identification (RFID) uses electromagnetic fields to identify and tra...
Catalog Number: (HACHLXV3229900001)
Supplier: Hach
Description: This Turbidity Suspended Solids (TSS) and sludge blanket level system can be used as an optimisation tool for monitoring process analyses or as an eas...
UOM: 1 * 1 items
Supplier: WTW
Description: The 'fine art' of measurement. Small but powerful: Optical finesse, reliable findings, ease of use and lab AQA with LIMS connection.
Supplier: Hach
Description: Portable turbidity meters designed for drinking water and industry applications.
Catalog Number: (LOVI194000)
Supplier: Lovibond
Description: The TurbiDirect is an autoranging turbidity meter designed for the laboratory or portable use.
UOM: 1 * 1 items
Catalog Number: (LOVI194300)
Supplier: Lovibond
Description: Ideal for field and environmental testing, the TB350 turbidity meter delivers the most reliable measurements for low range to high range samples with ...
UOM: 1 * 1 items
Supplier: EUTECH
Description: The turbidity meter operates on nephelometric principle for lab-accurate, highly repeatable results.
Catalog Number: (ORIOAQ3010)
Supplier: Thermo Orion
Description: The Orion AQUAfast AQ3010 is based on the nephelometric principle with 90° detection.
UOM: 1 * 1 items
Catalog Number: (ORIOAQ4500)
Supplier: Thermo Orion
Description: The ORION AQUAfast AQ4500 can provide the nephelometric and ratiometric principles of turbity measurement.
UOM: 1 * 1 items
Catalog Number: (707-0005)
Supplier: Hach
Description: The disc is designed to measure water transparency in situ via the semi-quantitative method.
UOM: 1 * 1 items
Catalog Number: (VELPR109B12150)
Supplier: VELP SCIENTIFIC
Description: The TB1 can measure the turbidity in aqueous samples for industrial and environmental fields.
UOM: 1 * 1 items
Catalog Number: (720-2686)
Supplier: LABONORD
Description: Measurement device of cell concentration as part of the process for liquid-based cytology for screening of cervical cancer as an alternative to the co...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom