You Searched For: Titrators
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SI Analytics
Description: TitroLine® 7000 automatic titrators are used in the laboratories of water supply companies, wastewater treatment plants, and the food and chemical ind...
Supplier: SI Analytics
Description: The TitroLine® 5000, successor to the TitroLine® easy, is a very easy to use automatic titrator for a variety of applications, including water/wastewa...
Supplier: SI Analytics
Description: The number of samples to be processed is growing constantly while at the same time the demands on reliability are increasing in accordance with GLP an...
Supplier: SI Analytics
Description: The Titronic® 300 is a motor-driven burette for manual titration and and a precise dosing instrument for dispensable liquids, solvents and titrating a...
Supplier: SI Analytics
Description: The Powerful TW 7450 developed for high sample throughput.
Catalog Number: (661-0246)
Supplier: Hach
Description: This automatic sample changer is designed to be used in analytical laboratories in connection with AT1000 Series titrators.
UOM: 1 * 1 items
Supplier: SI Analytics
Description: The headspace oven technique of the TO 7280 allows the separation of the water to be titrated from liquid, solid and semi-solid (paste-like) samples. ...
Supplier: SI Analytics
Description: These very compact, easy to operate instruments are ideal for coulometric water determination according to Karl Fischer in low water content, as well ...
Supplier: SI Analytics
Description: The Titroline® 7800 enhanced the universal features of the TitroLine® 7750 (potentiometric titration and volumetric KF titration), with an additional ...
Sale
Supplier: SI Analytics
Description: TitroLine® 7500 KF is a volumetric generalist for a wide range of uses. Applications include KF volumetry, dead-stop titrations (SO₂, bromine number),...
Supplier: SI Analytics
Description: The Titroline® 7750 is the all-rounder for both potentiometric titration and volumetric KF titration. The titrator combines the features of the potent...
Supplier: Hach
Description: Portable and robust digital Titrator for fast and precise titrations. High resolution (125 µl/digit) and high accuracy (±1%). Applications for more th...
Catalog Number: (661-0241)
Supplier: Mettler - Toledo
Description: This compact titrator is ideal for fast and precise potentiometric titrations. With its easy to use interface and space saving design, it is the optim...
UOM: 1 * 1 items
Catalog Number: (661-0210)
Supplier: Mettler - Toledo
Description: This volumetric KF titrator is ideal for fast and precise water content determination with easy operation. Samples with water content between 100 ppm ...
UOM: 1 * 1 items
Catalog Number: (HACH2270900)
Supplier: Hach
Description: This portable titrator enables analyses in the lab, the plant or the field.
UOM: 1 * 1 KIT
Supplier: Mettler - Toledo
Description: The C10S KF coulometric titrators are ideal for fast and precise water content determination with very simple operation. Samples with water content be...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom