Single channel electronic timer with memory, clock and certificate of calibration.
- Counts down and up from 23 hours, 59 minutes, 59 seconds
- 12/24 hour clock feature
- Separate hour, minute, second, start/stop and reset keys
- Last time set memory
- Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment
Certifications: Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Supplied with clip, stand, magnetic attachment, certificate of calibration, and one LR44 battery.