VWR®, Analogue Stopwatch
Supplier: VWR
VWR®
609-0132EA 128 EUR
609-0132
VWR®, Analogue Stopwatch
Timers Analogue Stopwatches
Crown stopper with frame ring.
  • Copper chromium plated casing
  • Separate side reset button
  • ¹/₅ second timing
  • Counts up to 30 minutes
W×D×H: 48×16×68 mm
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.