You Searched For: Timers
88  results were found
SearchResultCount:"88"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (609-0212)
Supplier: VWR Collection
Description: Quartz-crystal stopwatch with large LCD digits.
UOM: 1 * 1 items
Catalog Number: (609-0220)
Supplier: VWR Collection
Description: This three-channel lab timer is ideal for quality control and experimental requirements. It signals drying and incubating times, helps catch all test ...
UOM: 1 * 1 items
Catalog Number: (609-0195)
Supplier: VWR Collection
Description: Personalise with coloured faceplates.
UOM: 1 * 1 items
Catalog Number: (620-2220)
Supplier: VWR Collection
Description: This lab wall clock displays time, temperature, and humidity.
UOM: 1 * 1 items
Catalog Number: (609-0139)
Supplier: VWR Collection
Description: This electronic stopwatch includes a calendar and a clock.
UOM: 1 * 1 items
Catalog Number: (609-0205)
Supplier: VWR Collection
Description: This timer, with quartz-crystal accuracy, features count up and countdown functions.
UOM: 1 * 1 items
Catalog Number: (609-0225)
Supplier: VWR Collection
Description: This timer features a modern design, polished metal case and rubberised backing.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These digital timers allow the setting of four separate channels simultaneously in any combination of countdown or count-up (stopwatch).
Sale
Catalog Number: (609-0208)
Supplier: VWR Collection
Description: This timer is built in a white board with marker and eraser, which helps to keep track of time and other critical information. It provides different m...
UOM: 1 * 1 items
Catalog Number: (609-0184)
Supplier: VWR Collection
Description: Weight: 35,4 g.
UOM: 1 * 1 items
Catalog Number: (609-0231)
Supplier: VWR Collection
Description: View giant size digits on this bench timer from across the lab. Visible from shelves, in hoods, or perched on instruments. Eliminates 'squinting' to o...
UOM: 1 * 1 items
Catalog Number: (620-2300)
Supplier: VWR Collection
Description: View time, temperature, and humidity from anywhere.
UOM: 1 * 1 items
Catalog Number: (609-0213)
Supplier: VWR Collection
Description: Digital stopwatch, VWR®, Timing capacity: 10 h, No. of channels: 1, Accuracy: ±0,001 %, Black/blue, W×D×H: 54×19×66,7 mm, Memory: +
UOM: 1 * 1 items
Catalog Number: (609-0204)
Supplier: VWR Collection
Description: Digital timer, Three channel, VWR®, Timing capacity: 100 h, Up/down counting: ↕, No. of channels: 3, Accuracy: ±0,01 %, White/grey, W×D×H: 51×12,7×82,5 mm, With alarm: +, Memory: +
UOM: 1 * 1 items
Catalog Number: (609-0210)
Supplier: VWR Collection
Description: Digital timer, VWR®, Timing capacity: 100 min, Up/down counting: ↓, No. of channels: 1, Accuracy: ±0,01 %, Black, W×D×H: 63,5×12,7×63,5 mm, With alarm: +, Memory: +
UOM: 1 * 1 items
Catalog Number: (609-0197)
Supplier: VWR Collection
Description: Digital timer, One channel, VWR®, Timing capacity: 100 min, Up/down counting: ↕, No. of channels: 1, Accuracy: ±0,01 %, White/blue, W×D×H: 84×13×76 mm, With alarm: +, Memory: +
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 88
no targeter for Bottom