Lightweight, ergonomic overhead stirrer which is ideal for high viscosity mixing, large batch mixing or stirring products with medium viscosity. Microprocessor control provides feedback to the motor to maintain constant speed under changing viscosity, ensuring repeatable settings. The smooth start provides enhanced safety. VOS power control is particularly suitable for applications involving precise mixing documentation. It can be controlled using optional software and features integrated torque trend for viscosity control. An analogue interface enables the speed and torque to be recorded. Unit accommodates push-through stirring elements with shaft Ø up to 10 mm.
- Infinitely adjustable without gear shifting
- Microprocessor control
- Overload protection
- Digital speed display and RS232 interface
Certifications: IP protection class according to DIN EN 60529: IP 42
Delivery information: Supplied with chuck key and extension arm. Accessories must be ordered separately. Plate stand 431-2901 has slip resistant foil coating, a 16 mm diameter supporting rod and supports up to 5 kg.