VWR® VOS 14, Overhead Stirrer
Supplier: VWR
VWR®
441-0017EA 0 EUR
441-0017 441-0026
VWR® VOS 14, Overhead Stirrer
Stirrers Overhead Stirrers
This quiet, economical overhead stirrer is suitable for simple laboratory mixing, applications such as processing media with substantial temporary viscosity changes. Ideal for stirring substances with low to medium viscosities. Electronically controlled closed loop feedback to the motor to maintain constant speed, even when viscosity changes. The shaft accommodates stirring elements with shafts up to 8 mm in Ø.
  • Gearless, direct drive motor
  • Infinitely adjustable speed control
  • Easy to operate
  • Analogue speed display, overload protection
Certifications: IP protection class according to DIN EN 60529: IP 20
Delivery information: Supplied with chuck key and extension arm. Accessories must be ordered separately. Plate stand 431-2901 has slip resistant foil coating, a 16 mm diameter supporting rod and supports up to 5 kg. Please see website for details of accessories.