Multi-position magnetic hotplate stirrers, RT 5/10/15 power IKAMAG®
Supplier: IKA
IKAMAG®
442-2382EA 0 EUR
442-2382 442-2389 442-0001 442-0002 442-0003 IKAA2930305 442-2386
Multi-position magnetic hotplate stirrers, RT 5/10/15 power IKAMAG®
Stirrers Magnetic Hotplate Stirrers, Multi-position
High performance magnetic hotplate stirrers with 5, 10 or 15 stirring positions and integrated temperature control plate.
  • Space-saving design
  • Stirrers operate simultaneously
  • Ideal for series experiments due to precise and consistent temperature distribution
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
  • Temperature consistency in the medium ±2 K
Certifications: IP protection class according to DIN EN 60529: IP 42
Caution: Caution: Maximum media temperature (depending on the nature of the container) 70 °C.