These fluorimeters are available to cover a wide range of applications.
- Total Energy Transfer (TET) system for maximum sensitivity
- Intelligent Filter Modules (IFM) ensure correct filters are fitted
- Raw fluorescence and concentration values displayed
- Kinetic (timed reading) functions
- Time and date tag on stored reading enhances GLP
Ordering information: Interference filters are available from 250 to 700 nm (wavelength intervals typically 5 - 10 nm). For further details on specific applications and details of sample holder options, please contact your local VWR sales office.
Delivery information: Supplied with mains adapter for use on 90 to 230 V A/C supply with UK, EU and US mains leads and operating instructions. Emission and excitation filters are not included and should be purchased as separate items.