Vortex mixers, SA7 and SA8
Supplier: Cole Parmer
444-1601EA 394 EUR
444-1601 444-1602
Vortex mixers, SA7 and SA8
Shakers Vortex Shakers
For rapid mixing of samples in test tubes, small flasks and bottles. The SA8 can agitate microtitre plates with optional accessory 444-1603. Robust die-cast body avoids unwanted movement during use. Integral retort rod fixing allows vessels to be secured for safer mixing for long periods.
  • Ergonomic design
  • SA7 has fixed speed and automatic start when cap is depressed
  • SA8 has variable speed control and touch or continuous mode
  • BioCote® antimicrobial protection
IP protection class according to DIN EN 60529: IP 45
Delivery information: Optional accessory pack for use with SA8, 444-1603 includes: 1× plastic cradle for standard microtitre plate, 1× circular foam insert for beakers and flasks up to 500 ml, 1× rectangular insert (which holds 8×0,2, 8×0,5and 16×1,5/2,0 ml microcentrifuge tubes) and 1× solid rectangular foam insert for custom drilling.