You Searched For: Sieve Shakers
14  results were found
SearchResultCount:"14"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: HAVER & BOECKER
Description: These electromagnetically-driven test sieve shakers are designed for sieve analysis with optimum three-dimensional sieving action. The self-re-adjusti...
New Product
Supplier: HAVER & BOECKER
Description: The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a...
New Product
Supplier: HAVER & BOECKER
Description: For dry sieving. Tyler Ro-Tap® shakers RX 29 CE and RX 30 CE reproduce the circular and tapping motion of hand sieving, but in a repeatable mechanical...
New Product
Catalog Number: (510-1275)
Supplier: HAVER & BOECKER
Description: Using ultrasonic frequency variation for test sieving, even powders with separating cuts of ≤300 micron can be sieved. Fitting at least one HAVER-test...
UOM: 1 * 1 items
Catalog Number: (510-1379)
Supplier: Retsch GmbH
Description: The Retsch analytical air jet sieve shaker is particularly suitable for dispersion and deagglomeration of fine powders. Typical applications include c...
UOM: 1 * 1 items
Supplier: Retsch GmbH
Description: Retsch analytical sieve shakers are used in research and development, quality control of raw materials, interim and finished products, as well as in p...
Supplier: Retsch GmbH
Description: Analytical sieve shaker AS 200 tap is used in research and development, quality control of raw materials, interim and finished products, as well as in...
Catalog Number: (510-0048)
Supplier: Retsch GmbH
Description: With horizontal circular motion, for sieves 100/150/200/203/305 and 400 mm diameter.
UOM: 1 * 1 items
Catalog Number: (RETS30.026.0001)
Supplier: Retsch GmbH
Description: The analytical sieve shaker AS 450 control is used in research and development, quality control of raw materials, interim and finished products as wel...
UOM: 1 * 1 items
Supplier: ENDECOTTS
Description: Has been designed and manufactured to combine the advantages of low cost with the benefits of a well designed and engineered shaker incorporating many...
Catalog Number: (ENDEOCT200/23050)
Supplier: ENDECOTTS
Description: The Octagon 200 sieve shaker is suitable for sieving tasks both in the laboratory environment as well as onsite, where it provides optimum sieving act...
UOM: 1 * 1 items
Catalog Number: (510-4102)
Supplier: FRITSCH
Description: Analysette®3 spartanis a vibratory sieve shaker for all typical sieving tasks in the laboratory, with optical adjustment of the amplitude on the runni...
UOM: 1 * 1 items
Catalog Number: (510-4101)
Supplier: FRITSCH
Description: The high performance Analysette®3 Pro vibratory sieve shaker offers fast determination of quantitative particle size distribution in the laboratory. I...
UOM: 1 * 1 items
Catalog Number: (FRIT18.3020.00)
Supplier: FRITSCH
Description: The Analysette®18 heavy duty analytical sieve shaker was specially designed for large sieving quantities. It can effortlessly sieve up to 15 kg of mat...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 14 of 14
no targeter for Bottom