You Searched For: Rotating/Rocking Shakers
47  results were found
SearchResultCount:"47"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (444-0500)
Supplier: VWR Collection
Description: A unique design of rotator makes this unit suitable for a wide range of mixing applications in biochemistry, molecular biology and histochemistry. The...
UOM: 1 * 1 items
Catalog Number: (444-0148)
Supplier: VWR Collection
Description: The mini nutating mixer that combines the action of an orbital shaker and a rocker to produce a three-dimensional motion which provides gentle yet tho...
UOM: 1 * 1 items
Catalog Number: (700-0245)
Supplier: VWR Collection
Description: The Mini blot shaker combines the actions of a rocker and a shaker to produce a three-dimensional motion that is gentle enough for fragile gels and bl...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Rocking platform shakers have variable speed and adjustable tilt to provide optimum control of mixing conditions, a gentle wave motion is ideal for bl...
Catalog Number: (444-0756)
Supplier: VWR Collection
Description: Rocking platform shaker with low profile design can be used in most fume hoods and incubators. The cast aluminium base offers durability and stability...
UOM: 1 * 1 items
Catalog Number: (444-1343)
Supplier: VWR Collection
Description: The VWR® rotating mixer provides gentle to vigorous mixing of samples in up to 50 ml tubes.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Tube rollers are designed for the gentle, thorough mixing of a variety of containers, including Vacutainersâ„¢, bottles, test tubes, centrifuge tubes an...
Catalog Number: (444-0759)
Supplier: VWR Collection
Description: The low profile 3-D rotator waver provides precise speed control and a smooth, low foaming, three dimensional, wave motion. The cast aluminium base of...
UOM: 1 * 1 items
Supplier: IKA
Description: Shakers for smooth mixing tasks, 2D models feature a rocking motion and 3D units have three dimensional tumbling motion, basic models have fixed shaki...
Supplier: IKA
Description: Rotators for smooth yet effective mixing, the basic model has fixed speed, and the digital unit features infinitely adjustable speed. Ideal for biolog...
Catalog Number: (444-0932)
Supplier: GRANT INSTRUMENTS LTD.
Description: Compact rotator with easy to use controls, provides thorough end over end mixing of samples in tubes up to 50 ml. Applications include hybridisation r...
UOM: 1 * 1 items
Catalog Number: (444-1330)
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Platform shaker with rocking motion, designed for homogeneous mixing, ideal for flasks, media bottles and large vessels. Units have analogue dial cont...
UOM: 1 * 1 items
Supplier: IKA
Description: Roller shakers with 6 or 10 rolls, provide smooth rocking and rolling action, basic models have fixed speed, and the digital units feature infinitely ...
Catalog Number: (444-0927)
Supplier: GRANT INSTRUMENTS LTD.
Description: The sturdy PMR-100 variable speed and adjustable angle rocker is ideally suited for mixing solutions or nutrient media in vessels or bags. The unit pr...
UOM: 1 * 1 items
Catalog Number: (444-0353)
Supplier: GRANT INSTRUMENTS LTD.
Description: The PS-M3D is a fixed angle, variable speed, multifunction 3D rotator providing variable mixing options in a compact unit. Applications include immuno...
UOM: 1 * 1 items
Catalog Number: (444-0350)
Supplier: GRANT INSTRUMENTS LTD.
Description: The compact PS-3D variable speed, fixed angle 3D rotator is ideally suited for personal use. Applications include mixing in tubes, culture flasks, gel...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 47
no targeter for Bottom