You Searched For: Reciprocating Shakers
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (444-2909)
Supplier: VWR Collection
Description: Digital shaker with microprocessor control which is ideal for a range mixing applications, e.g. solubility studies and extraction procedures. Variable...
UOM: 1 * 1 items
Waste Reducing
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Promax shakers have a one dimensional back and forth motion for vigorous mixing of flasks and separating funnels in chemical applications, using analo...
Catalog Number: (442-1184)
Supplier: IKA
Description: Low profile laboratory shaker with digital display and a large platform. Continuous operation is guaranteed even under extreme loads. Suitable for ves...
UOM: 1 * 1 items
Supplier: IKA
Description: Compact flat shakers with electronic adjustment of speed and timer. Control model has an RS232/analogue interface and integrated end point positioning...
Supplier: Edmund Bühler GmbH
Description: Bench top reciprocating shakers which are suitable for continuous operation and accommodate loads of up to 30 kg. Universal tray accepts max. 11×1000 ...
Supplier: Edmund Bühler GmbH
Description: Reciprocating shakers, which are suitable for continuous or timer operation. The optional universal tray accepts a maximum of 5×1000 ml or 9×500&...
Catalog Number: (444-9422)
Supplier: Edmund Bühler GmbH
Description: Large, heavy duty reciprocating shaker for use as a floor standing unit only. Shaker is designed for continuous operation and accommodates loads of up...
UOM: 1 * 1 items
Catalog Number: (444-9120)
Supplier: Edmund Bühler GmbH
Description: Laboratory shaker which has functionality to change motion from orbital to reciprocating without using any tools. Sturdy, maintenance free shaker, sui...
UOM: 1 * 1 items
Catalog Number: (STUA51901-02)
Supplier: Cole Parmer
Description: Powerful yet quiet device is perfect for continuous use applications.
UOM: 1 * 1 items
Catalog Number: (OHAU30391830)
Supplier: OHAUS
Description: This digital reciprocating shaker is designed for a wide range of applications, including cell cultures, that require accurate and repeatable results....
UOM: 1 * 1 items
Supplier: WITEG LABORTECHNIK GMBH
Description: These digital reciprocating shakers with programmable digital feedback control system are ideal for culturing cells in hybridisation, staining and des...
Catalog Number: (444-0155)
Supplier: GERHARDT
Description: Laboratory scale platform shakers, which are ideal for extractions, microbiology application and environmental analysis. The units have a stable base,...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom